missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ RA Lamb 14-Drawer Slide Filing Cabinets, mini, grey

Vertically store up to 6000 76 x 26mm microscope slides with these 14-Drawer Cabinets.

Brand:  Epredia™ E43/12BLUE

 View more versions of this product

Product Code. 12131820

  • 500.00 € / Each
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Capacity of 3000 or 6000 slides depending on model
Specifications

Specifications

Slide Filing Cabinet
Blue
6000 Slides
76 mm
26 mm
6000
480 mm
480 mm
Steel
140 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.