Learn More
Epredia™ RA Lamb 14-Drawer Slide Filing Cabinets, mini, grey
Vertically store up to 6000 76 x 26mm microscope slides with these 14-Drawer Cabinets.
Brand:  Epredia™ E43/12GREY
 View more versions of this product
Product Code. 12151830
  • 429.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Capacity of 3000 or 6000 slides depending on model
Specifications
Specifications
Slide Filing Cabinet
Gray
6000 Slides
76 mm
26 mm
6000
480 mm
480 mm
Steel
140 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products