Learn More
Invitrogen™ Rabbit anti-Chicken IgY (H+L) Secondary Antibody, Biotin
Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen™ 31720
195.62 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31720 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31720 reacts with the heavy chains of chicken IgY and with the light chains common to most chicken immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Reconstitute with 1.5 mL of distilled water (1.3 mg/mL after restoration). Country of Origin: USA.
Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Chicken IgY (H+L) | |
Polyclonal | |
Biotin | |
Rabbit | |
1.5 mL | |
Secondary | |
4°C | |
Lyophilized |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
1.3 mg/mL | |
PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 | |
Affinity chromatography | |
RUO | |
Chicken | |
Secondary Antibody | |
IgG |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
109010I | Certificate of Analysis | 25/03/2025 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.