missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rabbit anti-Guinea Pig IgG (H+L) Secondary Antibody, HRP

Rabbit Polyclonal Secondary Antibody

Brand:  Invitrogen™ 614620

Product Code. 10671044

  • 292.00 € / 1mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Rabbit anti-Guinea Pig IgG (H+L) Secondary Antibody for Western Blot, IHC (P), IP, ELISA

Anti-Guinea Pig secondary antibodies are affinity-purified antibodies with well-characterized specificity for guinea pig immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Guinea Pig IgG (H+L)
Polyclonal
HRP
Rabbit
Affinity chromatography
RUO
Guinea Pig
Secondary Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, ELISA
1 mg/mL
PBS with 4mg/mL BSA, 40% glycerol and 0.19% Kathon™ CG; pH 7.4
Guinea Pig IgG.
1 mL
Secondary
4°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberAB411956 Certificate TypeCertificate of Analysis Date20/02/2025 Catalog Number
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Rabbit anti-Guinea Pig IgG (H+L) Secondary Antibody, HRP > 1 mL; HRP

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.