Learn More
MP Biomedicals™ Rabbit Serum
Sterile, Frozen Brown Solution. Combine with BSA to use as a blocking serum in immunocytochemistry and immunohistochemistry.
Brand:  MP Biomedicals™ 0219135780
 View more versions of this product
Product Code. 11480422
  • 65.93 € / 100mL
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
U.S. regulations
Rabbit
Rabbit Serum
4 to 7 g/dL
Membrane-filtered
6.5 to 8
-20°C
Liquid
100 mL
Sterile
Blocking agent in immunohistochemistry culture media component