missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Rabbit Serum

Sterile, Frozen Brown Solution. Combine with BSA to use as a blocking serum in immunocytochemistry and immunohistochemistry.

65.93 €

Specifications

Content And Storage -20°C
Species Rabbit
Form Liquid
Product Type Rabbit Serum
Concentration 4 to 7 g/dL
View More Specs

Products 1
Product Code Brand Concentration Form Quantity Price Quantity & Availability  
Product Code Brand Concentration Form Quantity Price Quantity & Availability  
11480422
View Documents
MP Biomedicals™
0219135780
4 to 7 g/dL Liquid 100 mL
65.93 €
100mL
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

-20°C
Liquid
4 to 7 g/dL
Membrane-filtered
6.5 to 8
Rabbit
Rabbit Serum
Sterile
Blocking agent in immunohistochemistry culture media component
Videos
SDS
Documents

Documents

Product Certifications