Learn More
Thermo Scientific™ Racks and Inserts for Refrigerated and Heated Bath Circulators
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
Brand:  Thermo Scientific™ 1600004
 View more versions of this product
Product Code. 11960715
  • 168.84 € / Each
Estimated Shipment: 01-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rack Insert; Includes top and bottom panels
Rack Insert
Stainless Steel
Bath Types A5B, A10B, A24B, S49, S19T, S14P, S21P
60 16 mm Tubes
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Racks and Inserts for Refrigerated and Heated Bath Circulators >