Learn More
Thermo Scientific™ Racks and Inserts for Refrigerated and Heated Bath Circulators
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
134.67 € - 249.24 €
Specifications
Material Stainless Steel
View More Specs
Products 12
Product Code Brand Description For Use With (Equipment) Holds Price Quantity & Availability  
Product Code Brand Description For Use With (Equipment) Holds Price Quantity & Availability  
10048294
View Documents
Thermo Scientific™
1600002
Stainless-steel rack Bath Types A5B, A10B, A24B, S49, S19T, S14P, S21P Depends on insert (purchase separately)
224.12 €
Each
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
11620961
View Documents
Thermo Scientific™
1600003
Rack Insert; Includes top and bottom panels Bath Types A5B, A10B, A24B, S49, S19T, S14P, S21P 100 10 mm Tubes
168.84 €
Each
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
11960715
View Documents
Thermo Scientific™
1600004
Rack Insert; Includes top and bottom panels Bath Types A5B, A10B, A24B, S49, S19T, S14P, S21P 60 16 mm Tubes
168.84 €
Each
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
10458434
View Documents
Thermo Scientific™
1600006
Rack Insert; Includes top and bottom panel with no holes Bath Types A5B, A10B, A24B, S49, S19T, S14P, S21P
148.74 €
Each
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
11977274
View Documents
Thermo Scientific™
1600079
Stainless-steel rack Bath Types A25B, A40, S21, S30 Depends on insert (purchase separately)
224.12 €
Each
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
11907284
View Documents
Thermo Scientific™
1600082
Rack Insert; Includes top and bottom panel Bath Types A25B, A40, S21, S30 13 25 mm Tubes
187.94 €
Pack of 2
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
11917284
View Documents
Thermo Scientific™
1600083
Rack Insert; Includes top and bottom panel with no holes Bath Types A25B, A40, S21, S30
134.67 €
Pack of 2
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
11680981
View Documents
Thermo Scientific™
1600026
Stainless-steel rack Bath Types S13, S12T, S15 Depends on insert (purchase separately)
249.24 €
Each
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
11927284
View Documents
Thermo Scientific™
1600084
Rack Insert; Includes top and bottom panels Bath Types S13, S12T, S15 39 10 mm Tubes
187.94 €
Pack of 2
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
11668008
View Documents
Thermo Scientific™
1600085
Rack Insert; Includes top and bottom panels Bath Types S13, S12T, S15 20 16 mm Tubes
187.94 €
Each
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
11947284
View Documents
Thermo Scientific™
1600086
Rack Insert; Includes top and bottom panels Bath Types S13, S12T, S15 8 25 mm Tubes
187.94 €
Pack of 2
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
11957284
View Documents
Thermo Scientific™
1600087
Rack Insert; Includes top and bottom panel with no holes Bath Types S13, S12T, S15
148.74 €
Pack of 2
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
Description
Description
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
Specifications
Specifications
Stainless Steel
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Racks and Inserts for Refrigerated and Heated Bath Circulators