Learn More
Fisherbrand™ Radiation Safety Carboy Covers
Idea cover for the protection of carboys and laboratory equipment.
Brand:  Fisherbrand™ CSR-COV
Product Code. 12307147
  • 723.00 € / Each
Estimated Shipment: 26-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For the protection of carboys and laboratory equipment
  • An open base allows the cover to be placed over the container or equipment
  • A hinged lid provides easy access
  • Can be used in conjunction with a laboratory safety tray
Specifications
Specifications
59 x 38 x 38cm (internal) and 60 x 40 x 40cm (external)
9 cm (internal) and 60 cm (external)
59cm
Radiation protection carboy cover
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Radiation Safety Carboy Covers > Dimensions: 38 x 38 x 59 cm (Internal); 40 x 40 x 40 (external); height: 590mm