missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RAPID™ Hp StAR™ Lateral Flow Kit

Rapidly detect Helicobacter pylori antigens in human stool samples with this lateral flow immunochromatographic assay.

Brand:  Thermo Scientific™ K671411-2

Product Code. 13275619

  • 1005.00 € / 20 tests

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

2°C to 8°C
H. pylori Antigen
Lateral Flow Kit
25 x wooden sticks, 20 x transparent test tubes, 20 x pasteur pipettes, 20 x test strips provided ready to use in a resealable foil pouch, sample diluent, instruction for use.
RAPID™ Hp StAR™ Lateral Flow Kit using Chromatographic Assay
Fecal
Enzyme Immunoassay Detection
20 Tests
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6174119 Certificate TypeCertificate of Analysis Date14/01/2025 Catalog NumberK671411-2
Lot Number6174118 Certificate TypeCertificate of Analysis Date21/11/2024 Catalog NumberK671411-2
Lot Number6174116 Certificate TypeCertificate of Analysis Date01/11/2024 Catalog NumberK671411-2
Lot Number6102715 Certificate TypeCertificate of Analysis Date16/09/2024 Catalog NumberK671411-2
Lot Number3675755 Certificate TypeCertificate of Analysis Date09/05/2024 Catalog NumberK671411-2
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ RAPID™ Hp StAR™ Lateral Flow Kit >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.