missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Rat BCRP Vesicles

Rat BCRP Vesicles are insect derived purified plasma membranes with inserted BCRP transport protein (Breast Cancer Resistance Protein, ABCG2).

Brand:  Gibco™ GM0007

Product Code. 10589710

  • 2335.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Frozen
Research Grade
Rat Vesicle
Vial
0.5 mL
Rat
Store at -80°C.
Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Rat BCRP Vesicles > 0.5 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.