Learn More
RBS™ Alkaline Cleaning Agent
Suitable for Soaking and Cleaning Surfaces and Radioactive Decontamination
284.00 € - 485.00 €
Specifications
Product Type Alkaline Cleaning Agent
Appearance Colorless
View More Specs
Products 3
Product Code Brand Product Line Price Quantity & Availability  
Product Code Brand Product Line Price Quantity & Availability  
12863512
View Documents
RBS
RBS50-DRUM
RBS 50 N/A N/A N/A
12823512
View Documents
RBS
RBS25-DRUM
RBS 25 N/A N/A N/A
12843512
View Documents
RBS
RBS35-DRUM
RBS 35
345.00 €
Each
Description
Description
  • RB525 is foaming, with a pH of 10.3 when using 20g per L of water, suitable for soaking
  • RB535 is foaming, with a pH of 10.3 when using 20g per L of water, suitable for soaking and permits lengthy cleaning without calcium precipitation
  • RB550 is non-foaming, with bacterial action with a pH of 10.6 when using 10g per L of water, suitable for soaking and automatic washing machines
  • All three are suitable for ultrasonic cleaners
  • RBSVIRO is a neutral foaming, surfacing, non-toxic, non-irritant and non-corrosive agent with a pH of 7, suitable for soaking
Recommended for use in research, pathology, chemistry, biology, food and drink, pharmaceutical, electronics, optical and glass industry laboratories.
Specifications
Specifications
Alkaline Cleaning Agent
Colorless
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
RBS™ Alkaline Cleaning Agent