Learn More
Thermo Scientific™ Reacti-Therm™ Magnetic Stirrers for Reacti-Vial™ Small Reaction Vials
Use these triangular stir bars for faster reaction times and smooth mixing in vials of the Thermo Scientific™ Reacti-Therm™ Derivatization System.
Brand:  Thermo Scientific™ TS-16010
189.46 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11597450
  • 190.41 € / Pack of 6
Explore more special offers
Description
Description
Use these triangular stir bars for faster reaction times and smooth mixing of small samples in conical-bottom vials of the Thermo Scientific™ Reacti-Therm™ Derivatization System. The stir bars provide simultaneous stirring of samples in multiple vials when used with a Reacti-Therm Heating/Stirring Module equipped with appropriate Reacti-Block Aluminum Blocks.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6 Pack
0.3 and 1mL Reacti-Vials
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Reacti-Therm™ Magnetic Stirrers for Reacti-Vial™ Small Reaction Vials > Fits 0.3 and 1mL Small Reacti-Vials
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.