missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Reagent Bottle, Narrow Neck, Soda-Lime Glass, Clear, Standard Taper Ground Joint Neck
Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
Description
A maximum usage temperature of +100°C is recommended. Thermal shock resistance 30 K. Hydrolytic class 3. Note on using DURAN™ glass stoppers with soda-lime glass bottles: If the bottle and the stopper have a temperature difference greater than 30°C, the glass stoppers can become stuck!
- All glass components
- Tight closure due to standard ground joint

Specifications
Specifications
Capacity (Metric) | 100 mL |
For Use With (Application) | Storage of air sensitive reagents, and chemicals; Storage of hygroscopic liquids, powders and granules |
Includes | With standard ground glass flat-headed stopper |
Material | Soda Lime Glass |
Quantity | 10/Pk. |
Autoclavable | Not Autoclavable |
Diameter (Metric) | 52 mm |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 7
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences Reagent Bottle, Narrow Neck, Soda-Lime Glass, Clear, Standard Taper Ground Joint Neck >
Spot an opportunity for improvement?Share a Content Correction