Learn More
Lenz Laborglasinstrumente™ Receiving Flask
Especially for use in rotary evaporators. Lenz Laborglasinstrumente™ Receiving Flasks are manufactured from durable DURAN™ glass.
Brand:  Lenz Laborglasinstrumente™ 03063573
 View more versions of this product
Product Code. 11826134
  • 93.00 € / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With spherical joint socket S 35
  • With inscription label
  • Available in a range of capacities
Specifications
Specifications
Duran Glass
Rotary Evaporators
166 mm
Receiving Flask
2000 mL