missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD14 Protein

A DNA sequence encoding the Homo sapiens (Human) CD14, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP2072-ec-100ug

 View more versions of this product

Product Code. 15975529

  • 554.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_000582.1
CD14 Protein
Research Use Only
CD14
Recombinant Protein
E. coli
TTPEPCELDD EDFRCVCNFS EPQPDWSEAF QCVSAVEVEI HAGGLNLEPF LKRVDADADP RQYADTVKAL RVRRLTVGAA QVPAQLLVGA LRVLAYSRLK ELTLEDLKIT GTMPPLPLEA TGLALSSLRL RNVSWATGRS WLAELQQWLK PGLKVLSIAQ AHSPAFSCEQ VRAFPALTSL DLSDNPGLGE RGLMAALCPH KFPAIQNLAL RNTGMETPTG VCAALAAAGV QPHSLDLSHN SLRATVNPSA PRCMWSSALN SLNLSFAGLE QVPKGLPAKL RVLDLSCNRL NRAPQPDELP EVDNLTLDGN PFLVPGTALP HEGSMN
Greater than 90% as determined by SDS-PAGE.
929
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers