missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD14 Protein
A DNA sequence encoding the Homo sapiens (Human) CD14, was expressed in the hosts and tags indicated.
160.00 € - 2485.00 €
Specifications
Accession Number | NP_000582.1 |
---|---|
Gene ID (Entrez) | 929 |
Name | CD14 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15985529
|
enQuireBio™
QP2072-ec-10ug |
10 μg | N/A | ||||||
15925539
|
enQuireBio™
QP2072-ec-50ug |
50 μg | N/A | ||||||
15975529
|
enQuireBio™
QP2072-ec-100ug |
100 μg |
554.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15905539
|
enQuireBio™
QP2072-ec-200ug |
200 μg | N/A | ||||||
15915539
|
enQuireBio™
QP2072-ec-500ug |
500 μg | N/A | ||||||
15995529
|
enQuireBio™
QP2072-ec-1mg |
1 mg |
2485.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_000582.1 | |
CD14 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
929 | |
Research Use Only | |
CD14 | |
Recombinant Protein | |
E. coli | |
TTPEPCELDD EDFRCVCNFS EPQPDWSEAF QCVSAVEVEI HAGGLNLEPF LKRVDADADP RQYADTVKAL RVRRLTVGAA QVPAQLLVGA LRVLAYSRLK ELTLEDLKIT GTMPPLPLEA TGLALSSLRL RNVSWATGRS WLAELQQWLK PGLKVLSIAQ AHSPAFSCEQ VRAFPALTSL DLSDNPGLGE RGLMAALCPH KFPAIQNLAL RNTGMETPTG VCAALAAAGV QPHSLDLSHN SLRATVNPSA PRCMWSSALN SLNLSFAGLE QVPKGLPAKL RVLDLSCNRL NRAPQPDELP EVDNLTLDGN PFLVPGTALP HEGSMN | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title