missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD14 Protein
A DNA sequence encoding the Homo sapiens (Human) CD14, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP2072-ec-10ug
This item is not returnable.
View return policy
Specifications
NP_000582.1 | |
CD14 Protein | |
Research Use Only | |
CD14 | |
Recombinant Protein | |
E. coli | |
TTPEPCELDD EDFRCVCNFS EPQPDWSEAF QCVSAVEVEI HAGGLNLEPF LKRVDADADP RQYADTVKAL RVRRLTVGAA QVPAQLLVGA LRVLAYSRLK ELTLEDLKIT GTMPPLPLEA TGLALSSLRL RNVSWATGRS WLAELQQWLK PGLKVLSIAQ AHSPAFSCEQ VRAFPALTSL DLSDNPGLGE RGLMAALCPH KFPAIQNLAL RNTGMETPTG VCAALAAAGV QPHSLDLSHN SLRATVNPSA PRCMWSSALN SLNLSFAGLE QVPKGLPAKL RVLDLSCNRL NRAPQPDELP EVDNLTLDGN PFLVPGTALP HEGSMN | |
Greater than 90% as determined by SDS-PAGE. |
929 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction