missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Complement C3 Protein
A DNA sequence encoding the Homo sapiens (Human) Complement C3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8695-ec-1mg
This item is not returnable.
View return policy
Specifications
NP_000055.2 | |
Complement C3 Protein | |
Research Use Only | |
C3 | |
Recombinant Protein | |
E. coli | |
YSIITPNILR LESEETMVLE AHDAQGDVPV TVTVHDFPGK KLVLSSEKTV LTPATNHMGN VTFTIPANRE FKSEKGRNKF VTVQATFGTQ VVEKVVLVSL QSGYLFIQTD KTIYTPGSTV LYRIFTVNHK LLPVGRTVMV NIENPEGIPV KQDSLSSQNQ LGVLPLSWDI PELVNMGQWK IRAYYENSPQ QVFSTEFEVK | |
Greater than 90% as determined by SDS-PAGE. |
718 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction