missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Complement C3 Protein
A DNA sequence encoding the Homo sapiens (Human) Complement C3, was expressed in the hosts and tags indicated.
160.00 € - 2485.00 €
Specifications
Accession Number | NP_000055.2 |
---|---|
Gene ID (Entrez) | 718 |
Name | Complement C3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16016001
|
enQuireBio™
QP8695-ec-10ug |
10 μg | N/A | ||||||
16056001
|
enQuireBio™
QP8695-ec-50ug |
50 μg | N/A | ||||||
16006001
|
enQuireBio™
QP8695-ec-100ug |
100 μg |
554.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16036001
|
enQuireBio™
QP8695-ec-200ug |
200 μg | N/A | ||||||
16046001
|
enQuireBio™
QP8695-ec-500ug |
500 μg | N/A | ||||||
16026001
|
enQuireBio™
QP8695-ec-1mg |
1 mg |
2485.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_000055.2 | |
Complement C3 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
718 | |
Research Use Only | |
C3 | |
Recombinant Protein | |
E. coli | |
YSIITPNILR LESEETMVLE AHDAQGDVPV TVTVHDFPGK KLVLSSEKTV LTPATNHMGN VTFTIPANRE FKSEKGRNKF VTVQATFGTQ VVEKVVLVSL QSGYLFIQTD KTIYTPGSTV LYRIFTVNHK LLPVGRTVMV NIENPEGIPV KQDSLSSQNQ LGVLPLSWDI PELVNMGQWK IRAYYENSPQ QVFSTEFEVK | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title