missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IL-22 (HEK293-expressed) Protein
Brand: R&D Systems™ 11311-IL-01M
This item is not returnable.
View return policy
Description
Measured by its ability to induce IL-10 secretion in COLO 205 human colorectal adenocarcinoma cells. Nagalakshmi, M.L. et al. (2004) International Immunopharmacology 4:679. The ED50 for this effect is 0.160-1.60 ng/mL.
Specifications
Q9GZX6.1 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
M.W. Observed: 28-32 kDa, under reducing conditions., M.W. Theoretical: 17 kDa | |
Human embryonic kidney cell, HEK293-derived human IL-22 protein Ala34-Ile179 | |
<0.10 EU per 1μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Bioactivity | |
50616 | |
1 mg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
Cytokine Zcyto18, IL-10-related T-cell-derived inducible factor, IL22, IL-D110, IL-TIF, ILTIFIL-10-related T-cell-derived-inducible factor, IL-TIFMGC79382, interleukin 22, interleukin-22, MGC79384, TIFa, TIFIL-23, zcyto18 | |
Reconstitute the 10 μg size at 100 μg/mL in PBS. Reconstitute all other sizes at 500 μg/mL in PBS. | |
IL-22 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction