Learn More
Novus Biologicals™ Recombinant Human KLHDC2 GST (N-Term) Protein
Recombinant Protein
Brand: Novus Biologicals™ H00023588-P01-2ug
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifications
Western Blot, ELISA, Protein Array, Immunoaffinity Purification | |
23588 | |
2 μg | |
HCA33, HCLP-1Host cell factor-like protein 1, Hepatocellular carcinoma-associated antigen 33, Host cell factor homolog LCP, kelch domain containing 2, kelch domain-containing protein 2, LCP | |
>80% by SDS-PAGE and Coomassie blue staining |
50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. | |
MolecularWeight-theroretical: 72.5 kDa | |
Store at -80°C. Avoid freeze-thaw cycles. | |
KLHDC2 | |
KLHDC2 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.