missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human P-Selectin/CD62P Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ ADP3-050
311.87 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human P-Selectin/CD62P Protein is derived from CHO. The Recombinant Human P-Selectin/CD62P Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Sorbitol. | |
80 kDa | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
6403 | |
P-Selectin/CD62P | |
Chinese Hamster Ovary cell line,CHO-derived human P-Selectin/CD62P protein Trp42-Ala771 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |