missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human P-Selectin/CD62P Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
408.00 € - 1045.00 €
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Sorbitol. |
---|---|
Gene ID (Entrez) | 6403 |
Molecular Weight (g/mol) | 80 kDa |
Name | P-Selectin/CD62P |
Source | Chinese Hamster Ovary cell line,CHO-derived human P-Selectin/CD62P protein Trp42-Ala771 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18197430
|
R&D Systems™
ADP3-200 |
200 μg |
1045.00 €
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18188480
|
R&D Systems™
ADP3-050 |
50 μg |
408.00 €
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human P-Selectin/CD62P Protein is derived from CHO. The Recombinant Human P-Selectin/CD62P Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Sorbitol. | |
80 kDa | |
Chinese Hamster Ovary cell line,CHO-derived human P-Selectin/CD62P protein Trp42-Ala771 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
6403 | |
P-Selectin/CD62P | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |