Learn More
R&D Systems™ Recombinant Mouse R-Spondin 1 (CHO-expressed) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 7150-RS-025
279.90 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18361418
  • 367.00 € / 25µg
Explore more special offers
Description
Description
The Recombinant Mouse R-Spondin 1 (CHO-expressed) Protein is derived from CHO. The Recombinant Mouse R-Spondin 1 (CHO-expressed) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
27 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
192199
R-Spondin 1
Chinese Hamster Ovary cell line,CHO-derived mouse R-Spondin 1 protein Met1-Gln265
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.