missing translation for 'onlineSavingsMsg'
Learn More

Novex™ Recombinant Protein G - Sepharose™ 4B

Recombinant Protein G - Sepharose™ 4B

524.00 € - 2865.00 €

Specifications

Product Type Protein G Sepharose
Product Line Sepharose™
Column Type Affinity
Stationary Phase Protein G
Form Suspension
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10573395
View Documents
Novex™
101242
5 mL
974.00 €
5mL
Please to purchase this item. Need a web account? Register with us today!
10078792
View Documents
Novex™
101241
2 mL
524.00 €
2mL
Please to purchase this item. Need a web account? Register with us today!
10405533
View Documents
Novex™
101243
25 mL
2865.00 €
25mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Recombinant protein G is produced in a strain of E. coli which contains the gene for protein G from Streptococcus sp. This product is prepared by covalently coupling highly purified rec-Protein G to Sepharose™ 4B. It is supplied as a 50% suspension containing rec-Protein G-Sepharose™ 4B + PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.1% Proclin).

Applications: Immunoglobulin affinity purification and Immunoprecipitation.

Recombinant protein G binds to the Fc region of many types of immunoglobulin G including:

  • all four subclasses of human IgG
  • mouse IgG
  • rabbit IgG
  • guinea pig IgG
  • rat IgG
  • goat IgG
  • sheep IgG
  • cow IgG
  • horse IgG

    Recombinant protein G does not bind to IgG from chicken or cat, or to human IgA, IgM, or serum albumin. It binds weakly to IgG from dog.
  • Specifications

    Specifications

    Protein G Sepharose
    Affinity
    Suspension
    Sepharose™
    Protein G
    Videos
    SDS
    Documents

    Documents

    Product Certifications

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.