Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O Polyvalent
Used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.
Brand:  Thermo Scientific™ R30858201
 View more versions of this product
Product Code. 11993322
  • 158.00 € / 2mL
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Remel Agglutinating Sera, Salmonella O is used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.
  • Select sera also for use as positive controls for stained Salmonella suspensions
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Positive
Salmonella O Antigen
Polyvalent
2 mL/Vial
Salmonella O
Salmonella O Polyvalent (group A - S)
Agglutinating Sera
Group A-S
2 mL