missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O Polyvalent
Used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.
146.00 € - 151.00 €
Specifications
Control Sets | Positive |
---|---|
Detectable Analytes | Salmonella O Antigen |
Type | Agglutinating Sera |
Material | Polyvalent |
Packaging Type | 2 mL/Vial |
Product Code | Brand | Description | Detectable Analytes | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Detectable Analytes | Quantity | Price | Quantity & Availability | |||
12627499
|
Thermo Scientific™
R30858101 |
Salmonella O Polyvalent (group A - G) | Salmonella O Antigen | 2 mL |
151.00 €
Each |
Estimated Shipment: 27-09-2024
Log in to see stock. |
|||
11993322
|
Thermo Scientific™
R30858201 |
Salmonella O Polyvalent (group A - S) | Salmonella O Antigen | 2 mL |
146.00 €
2mL |
Estimated Shipment: 26-09-2024
Log in to see stock. |
|||
Description
Remel Agglutinating Sera, Salmonella O is used for identification of somatic O antigens of Salmonella strains by slide and/or tube agglutination test.
- Select sera also for use as positive controls for stained Salmonella suspensions
Specifications
Positive | |
Agglutinating Sera | |
2 mL/Vial | |
Salmonella O |
Salmonella O Antigen | |
Polyvalent | |
2 mL |