Learn More
Thermo Scientific™ Agglutinating Sera, Salmonella O
Serologically identify somatic O antigens of Salmonella strains by slide agglutination test using Agglutinating Sera, Salmonella O.
Brand: Thermo Scientific™ R30957601
Description
Serologically identify somatic O antigens of Salmonella strains by slide agglutination test for epidemiological and diagnostic purposes using Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O. The sera are absorbed to remove agglutinins for other Salmonella antigens and the paracolon α agglutinin. Salmonella Somatic Agglutinating Sera (ZC series) are intended for the identification of O antigens.
- Select sera also for use as positive controls for stained Salmonella suspensions

Specifications
Positive | |
Salmonella O Antigen | |
Group F | |
2 mL |
Salmonella O Factor 11 (group F) | |
Agglutinating Sera | |
2 mL/Vial | |
Salmonella O |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6125072 | Certificate of Analysis | 09/09/2024 | R30957601 |
3775693 | Certificate of Analysis | 18/03/2024 | R30957601 |
3623663 | Certificate of Analysis | 20/06/2023 | R30957601 |
3446172 | Certificate of Analysis | 07/09/2022 | R30957601 |
3337376 | Certificate of Analysis | 08/10/2021 | R30957601 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.