missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Agglutinating Sera, Salmonella O

Serologically identify somatic O antigens of Salmonella strains by slide agglutination test using Agglutinating Sera, Salmonella O.

147.00 € - 163.00 €

Specifications

Control Sets Positive
Detectable Analytes Salmonella O Antigen
Type Agglutinating Sera
Packaging Type 2 mL/Vial
Quantity 2 mL
View More Specs

Products 11
Product Code Brand Description Detectable Analytes Quantity Price Quantity & Availability  
Product Code Brand Description Detectable Analytes Quantity Price Quantity & Availability  
11933332
View Documents
Thermo Scientific™
R30956801
Salmonella O Factors 3, 10, 15, 19 (group E) Salmonella O Antigen 2 mL
163.00 €
2mL
Please to purchase this item. Need a web account? Register with us today!
12653638
View Documents
Thermo Scientific™
R30958001
Salmonella O Factor 16 (group I) Salmonella O Antigen 2 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12697559
View Documents
Thermo Scientific™
R30957601
Salmonella O Factor 11 (group F) Salmonella O Antigen 2 mL
163.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
12663648
View Documents
Thermo Scientific™
R30959101
Salmonella O Factor 39 (group Q) Salmonella O Antigen 2 mL
163.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
12697569
View Documents
Thermo Scientific™
R30958601
Salmonella O Factor 27 (group B) Salmonella O Antigen 2 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12687559
View Documents
Thermo Scientific™
R30957501
Salmonella O Factor 10 (group E1) Salmonella O Antigen 2 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12623638
View Documents
Thermo Scientific™
R30957701
Salmonella O Factor 13, 22 (group G) Salmonella O Antigen 2 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12643638
View Documents
Thermo Scientific™
R30957901
Salmonella O Factor 15 (group E2) Salmonella O Antigen 2 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12633638
View Documents
Thermo Scientific™
R30957801
Salmonella O Factor 14 (group H) Salmonella O Antigen 2 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12683638
View Documents
Thermo Scientific™
R30958301
Salmonella O Factor 19 (group E4) Salmonella O Antigen 2 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11944092
View Documents
Thermo Scientific™
R30957301
Salmonella O Factor 9 (group D) Salmonella O Antigen 2 mL
163.00 €
2mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Serologically identify somatic O antigens of Salmonella strains by slide agglutination test for epidemiological and diagnostic purposes using Thermo Scientific™ Remel™ Agglutinating Sera, Salmonella O. The sera are absorbed to remove agglutinins for other Salmonella antigens and the paracolon α agglutinin. Salmonella Somatic Agglutinating Sera (ZC series) are intended for the identification of O antigens.

Serological tests are based on the fact that antibodies in serum, produced in response to exposure to bacterial antigens, will agglutinate with bacteria carrying homologous antigens.

Serologically identify somatic O antigens of Salmonella strains by slide and/or tube agglutination tests for epidemiological and diagnostic purposes using Agglutinating Sera, Salmonella O.

  • Rapid: Results within 1 minute.
  • Simple, ready to use: Makes the procedure easy to implement and run

The genus Salmonella is classified in the Kauffmann-White Scheme into serotypes according to combinations of somatic (O) and flagellar (H) antigens, which are identified by agglutination tests1. Agglutinating sera should be used in confirmatory tests but may also be used with appropriate caution in screening tests2. These simple tests rapidly detect specific bacterial antigens for the identification, grouping and/or typing of Salmonella and other food-borne microorganisms. This incorporates agglutinating sera for the identification somatic O antigens of Salmonella (groups A-S), allowing the fast, accurate and reliable identification of Salmonella strains.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Specifications

Positive
Agglutinating Sera
2 mL
Salmonella O Antigen
2 mL/Vial
Salmonella O
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Agglutinating Sera, Salmonella O

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.