missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Shigella boydii Polyvalent
Used for serological identification of Shigella cultures by rapid slide agglutination test.
Brand: Thermo Scientific™ R30164101
This item is not returnable.
View return policy
Description
Remel Agglutinating Sera, Shigella boydii Polyvalent are used for serological identification of Shigella cultures by rapid slide agglutination test.
Specifications
2°C to 8°C | |
Yes | |
Vial | |
2 mL |
Shigella boydii Polyvalent 3 (12 to 15) | |
Shigella | |
2 mL/Vial |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction