Learn More
Thermo Scientific™ Remel™ McFarland Equivalence Turbidity Standards
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
Brand:  Thermo Scientific™ R20421
456.67 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10036782
Description
Description
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
  • Tube size is 15 x 103mm and fits in most spectrophotometers
  • Polystyrene technology allows longer shelf life
Includes:
1 each of 0.5, 1.0, 2.0, 3.0, 4.0.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Turbidity Standard Set (1 each of 0.5, 1.0, 2.0, 3.0, 4.0)
1 each of 0.5, 1.0, 2.0, 3.0, 4.0
15 x 103 mm