Learn More
Thermo Scientific™ PathoDx™ Respiratory Virus Panel Influenza A Reagent
PathoDx Respiratory Virus Panel Influenza A Reagent along with RVP kit is a direct immunofluorescence test used to detect Influenza A.
Brand: Thermo Scientific™ R62405
Description
Use Thermo Scientific™ Remel™ PathoDx™ Respiratory Virus Panel Influenza A Reagent along with PathoDx™ Respiratory Virus Panel (RVP) kit to perform a direct immunofluorescence test for the qualitative detection of Influenza A in prepared patient specimens following growth in cell culture. Influenza viruses are negative-stranded RNA viruses of the Orthomyxoviridae family. Influenza infections are associated with seasonal outbreaks of respiratory disease, such as pharyngitis, croup, bronchitis, and pneumonia1. Usually one type, either type A or B, will predominate during each epidemic2.
Influenza A comprises of a single genus and shows antigenic variation in the hemagglutinin and neuraminidase proteins. Influenza A subtypes are identified by the hemagglutinin and neuraminidase antigens, H1-H13 and N1-N9, respectively3. Strains H1N1, H3N2 have been associated with pandemics in humans3.
- Ready-to-use – Prepared reagents
- Easy-to-store – Store at 2-8°C
- Easy to interpret – Fluorescence is nuclear, cytoplasmic, or both. Nuclear staining is uniformly bright, and cytoplasmic staining is often punctated with large inclusions
Influenzae A Reagent (Blue cap) is one dropper bottle containing 2.0 mL (approximately 40 tests) of fluorescein-labeled purified specific murine monoclonal antibodies, and Evans blue counterstain in a proteinstabilized buffer solution, with 0.098% sodium azide as preservative. Influenza A can be isolated in MRC-5, RD, A549, HL, and MDCK tube or shell vial cultures with or without the presence of trypsin3,1.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications
PathoDx™ Respiratory Virus Panel Influenzae A Reagent | |
Nasopharyngeal | |
1 x 2 mL |
Influenza | |
Reagent |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
852013-COA | Certificate of Analysis | 17/04/2017 | R62405 |
1434934-COA | Certificate of Analysis | 15/04/2017 | R62405 |
1198130-COA | Certificate of Analysis | 15/04/2017 | R62405 |
821560-COA | Certificate of Analysis | 15/04/2017 | R62405 |
1017372-COA | Certificate of Analysis | 15/04/2017 | R62405 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.