Learn More
Gilson™ REPET-TIPS™ Repeater Pipette Tips
GREENER_CHOICE
Specifically made for the Gilson™ REPETMAN™ positive-displacement electronic pipette, REPET-TIPS™ cover a range of volumes and are recognized by the REPETMAN™ operating system when placed on the pipette.
291.00 €
Specifications
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Model REPET-TIP™
View More Specs
Products 2
Product Code Brand Volume (Metric) Graduations Price Quantity & Availability  
Product Code Brand Volume (Metric) Graduations Price Quantity & Availability  
15344785
View Documents
Gilson™
F164528
1 mL 20 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15354785
GREENER_CHOICE
View Documents
Gilson™
F164558
10 mL 200 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Tip-size encoding system of the REPETMAN™ automatically identifies the REPET-TIP™ attached and displays the appropriate volume on the LCD screen
  • Made of polypropylene
  • Features volume appropriate graduations for precise measurement
Specifications
Specifications
Non-filtered
Clear
REPET-TIP™
66 mm
Yes
Hinged Rack
Repeater Pipette Tips
Single-use
Yes
Polypropylene
Sterile
Not Autoclavable
2.5 in
ISO 9001, ISO 17025, ISO 8655
Pipetting
100 Tips
Clear
Gilson Repetman
REPET-TipS
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Gilson™ REPET-TIPS™ Repeater Pipette Tips