Learn More
Research Octane Number Standard ASTM D 2699 (Nominal value: 98), Paragon Scientific™
Fuels Testing Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for Octane Number (Research Method) ASTM D2699
Brand: Paragon Scientific CRM-ORGA
Description
- Manufactured in accordance with ASTM / EN or IP test methods
- Certified by Round Robin Method by independant test laboratories
- Ensures good interlaboratory correlation
- Supplied in tamper evident UN compliant security pack
- Pack size 1000mL
- 12 months shelf life
Specifications
1 L | |
UN 1203 |
Safety and Handling
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Carcinogenicity Category 1B
- Flammable liquid Category 2
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H361-Suspected of damaging fertility or the unborn child.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P370+P378-In case of fire: Use dry chemical, CO2, water spray or alcohol-resistant foam to extinguish.
- P391-Collect spillage.
- P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
- MIXTURE LIST-Contains : Gasoline : low boiling point naphtha
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Nominal values quoted. Certified Values will vary from batch to batch.