missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Blot Module for MiniVE Mini Vertical Electrophoresis Unit

Can be used to transfer two minigels inside the MiniVE tank

Brand:  Hoefer™ SE302

Product Code. 11985418

  • 599.00 € / Each
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Add to basket

Includes: Blot module, Three 0.25 in. (6 mm) thick Dacron™ sponges, 25 Sheets of blotter paper
This item is not returnable. View return policy

Description

Description

  • MiniVE accommodates up to two blot modules simultaneously
  • Four gels can be transferred in 45 min. with only 300mL buffer
Specifications

Specifications

EN61010-1, UL61010A-1, CSA C22.2 1010.1, CE
Blot Module
Blot module, 25 sheets blotter Paper, four 6mm thick Dacron™ sponge
45°C
300 mL
10.5 or 8 cm
10 cm
100 mA
10 x 10.5 cm or 10 x 8 cm Precast Gel
Up to 80%
113°F
10.14 oz.
4.13 or 3.14 in.
3.93 in.
15 w
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hoefer™ Blot Module for MiniVE Mini Vertical Electrophoresis Unit > 10 x 10.5cm or 10 x 8cm Gel Size

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.