Learn More
R&L Enterprises™ Retort Stand Clamps
Enables articles from 2 to 90mm in diameter to be clamped securely. R&L Enterprises™ Retort Stand Clamps feature aluminum alloy jaws with other parts of plated brass.
133.72 € - 141.44 €
Specifications
Quantity 5/Pk.
Type Clamp Retort Stand
Material Aluminum Alloy, Brass
For Use With (Equipment) For use with 2 to 90mm diameter
Jaw Type Cork Lined
View More Specs
Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
11517722
View accessory products View Documents
R&L Enterprises™
9505
Metal headed clamping screw
133.72 €
Pack of 5
In Stock
to see stock.
Edge
Add to basket
11888203
View Documents
R&L Enterprises™
9505/P
Plastic headed clamping screw
141.44 €
Pack of 5
In Stock
to see stock.
Edge
Add to basket
Description
Description
Enables articles from 2 to 90mm in diameter to be clamped securely. R&L Enterprises™ Retort Stand Clamps feature aluminum alloy jaws with other parts of plated brass.
  • Design ensures a firm grip at all angles of opening
  • Cork liners for the jaws are well secured
  • Available with either metal or plastic clamping screw head
Note:
The clamp and boss head are sold separately.
Specifications
Specifications
5/Pk.
Aluminum Alloy, Brass
Cork Lined
Clamp Retort Stand
For use with 2 to 90mm diameter
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
R&L Enterprises™ Retort Stand Clamps