Learn More
R&L Enterprises™ Retort Stand Clamps
Enables articles from 2 to 90mm in diameter to be clamped securely. R&L Enterprises™ Retort Stand Clamps feature aluminum alloy jaws with other parts of plated brass.
Brand:  R&L Enterprises™ 9505
 View more versions of this product
Product Code. 11517722
  • 133.72 € / Pack of 5
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Includes:
Clamp only. (Boss head is sold separately.)
Description
Description
Enables articles from 2 to 90mm in diameter to be clamped securely. R&L Enterprises™ Retort Stand Clamps feature aluminum alloy jaws with other parts of plated brass.
  • Design ensures a firm grip at all angles of opening
  • Cork liners for the jaws are well secured
  • Available with either metal or plastic clamping screw head
Note:
The clamp and boss head are sold separately.
Specifications
Specifications
5/Pk.
Aluminum Alloy, Brass
For use with 2 to 90mm diameter
Clamp Retort Stand
Metal headed clamping screw
Cork Lined
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
R&L Enterprises™ Retort Stand Clamps > Includes: Metal headed clamping screw