missing translation for 'onlineSavingsMsg'
Learn More

RETSCH CryoMill Grinding Jar

Helps provide efficient milling

Brand:  RETSCH 01.462.0290

Product Code. 10247954

  • 1035.00 € / Each
Estimated Shipment: 16-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stainless-steel grinding jar with screw-top design
  • Grinding jar is continually cooled with liquid nitrogen from the integrated cooling system before and during the grinding process

Specifications

Specifications

Grinding Jar
5 mL
Ideal for cryogenic grinding
Stainless Steel
0.16 oz.
Screw Top
Adapter of CryoMill cryogenic grinder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
RETSCH CryoMill Grinding Jar > 5mL, Stainless-steel Grinding Jar

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.