missing translation for 'onlineSavingsMsg'
Learn More

RETSCH Sieve Shaker AS 200 Basic

Use this sieve shaker in research & development, quality control of raw materials, interim and finished products as well as in production monitoring. Retsch™ Sieve Shaker AS 200 features digital setting and display of performance and time.

Brand:  RETSCH 30.030.0001

Product Code. 15562515

  • 2995.00 € / Each
Estimated Shipment: 20-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • NEW: digital display of performance and time
  • Suitable for dry and wet sieving
  • Excellent separation efficiency even with short sieving times
  • Efficient electromagnetic drive
  • 3-D throwing motion which ensures optimum use of the open sieve area and lets the sample move Equally over the whole sieving surface
  • Sieve stack up to 510 mm height
  • Free adjustment of all process parameters (time, performance)
  • Optional comfort clamping device
  • Optional evaluation software EasySieve
  • Maintenance-free
  • Easy operation, ergonomic design
  • Standard delivery without clamping device, this must be ordered separately
Specifications

Specifications

3 kg
Sieve Shaker
384 mm
417 mm
Digital, 1 to 99 min.
230 V
50 Hz
212 mm
Retsch Sieve Shaker AS 200 Basic
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
RETSCH Sieve Shaker AS 200 Basic > Sieve Shaker

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.