Learn More
RETSCH Test Sieves - 14 Mesh or Greater
Stainless steel with wire gauze
Brand:  RETSCH 60.150.000212
 View more versions of this product
Product Code. 10752003
  • 134.00 € / Each
Estimated Shipment: 22-04-2025
to see stock.
Explore more special offers
Add to basket
Specifications
2 in.
212 μm
ASTM E11
Stainless Steel
No. 70
8 in.
5.08 cm
Test Sieve
8 in.
70 Mesh
Ideal for cement clinker, chemicals, coffee, construction materials, fertilizers, fillers, flours, grains, Metals powders, Minerals, nuts, Plastics, sand, seeds, soils, washing powder etc.
Stainless Steel