Learn More
RETSCH ZM 200 Mill Accessories
Push-fit assembly, with wear-resistant coating
Brand:  RETSCH 02.608.0044
4161.00 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10237940
  • 4490.00 € / Each
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
For use with ZM 200 Mixer Mill. Choose rotor based on properties of sample, required final fineness and subsequent analysis.
  • Stainless-steel, push-fit, with wear-resistant coating
  • 6-tooth rotor for coarse, bulky, fibrous goods such as feed pellets, hay and straw
  • 12-tooth rotor for medium-coarse goods such as wheat, oats, corn, tablets, powder coatings and plastics
Specifications
Specifications
12-tooth Rotor
Ultra Centrifugal mill ZM 200
Wear-resistant coating
Stainless Steel