Learn More
RETSCH ZM 200 Mill Accessories
Push-fit assembly, with wear-resistant coating
4490.00 €
Specifications
For Use With (Equipment) Ultra Centrifugal mill ZM 200
Includes Wear-resistant coating
Material Stainless Steel
View More Specs
Products 2
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
10237940
View Documents
RETSCH
02.608.0044
12-tooth Rotor
4490.00 €
Each
Estimated Shipment: 20-03-2025
to see stock.
Add to basket
10370201
View Documents
RETSCH
02.608.0043
6-tooth Rotor
4490.00 €
Each
Estimated Shipment: 20-03-2025
to see stock.
Add to basket
Description
Description
For use with ZM 200 Mixer Mill. Choose rotor based on properties of sample, required final fineness and subsequent analysis.
  • Stainless-steel, push-fit, with wear-resistant coating
  • 6-tooth rotor for coarse, bulky, fibrous goods such as feed pellets, hay and straw
  • 12-tooth rotor for medium-coarse goods such as wheat, oats, corn, tablets, powder coatings and plastics
Specifications
Specifications
Ultra Centrifugal mill ZM 200
Wear-resistant coating
Stainless Steel