Learn More
RETSCH ZM 200 Mill Accessories
Push-fit assembly, with wear-resistant coating
Brand: RETSCH 02.608.0043
4161.00 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
For use with ZM 200 Mixer Mill. Choose rotor based on properties of sample, required final fineness and subsequent analysis.
- Stainless-steel, push-fit, with wear-resistant coating
- 6-tooth rotor for coarse, bulky, fibrous goods such as feed pellets, hay and straw
- 12-tooth rotor for medium-coarse goods such as wheat, oats, corn, tablets, powder coatings and plastics
Specifications
6-tooth Rotor | |
Ultra Centrifugal mill ZM 200 | |
Wear-resistant coating | |
Stainless Steel |