missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
11377161
|
Thermo Scientific™
22.0087 |
Reverse Osmosis membrane |
149.75 €
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
11830820
|
Thermo Scientific™
D9006 |
Replacement Reverse Osmosis membrane for ROpure Infinity Systems |
2410.00 €
Each |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||||
11377131
|
Thermo Scientific™
09.2003 |
Reverse Osmosis membrane with integrated pretreatment, 3L/hr |
364.82 €
Each |
In Stock
Log in to see stock. ![]() |
||||||
11327141
|
Thermo Scientific™
09.2006 |
Reverse Osmosis membrane with integrated pretreatment, 6L/hr |
436.17 €
Each |
In Stock
Log in to see stock. |
||||||
11337161
|
Thermo Scientific™
22.0046 |
Reverse Osmosis membrane, for use with LabTower EDI 15, LabTower RO 20/60, LabTower TII 20/60, Pacific RO 3/7/12/20, Pacific TII 3/7/12/20 |
149.75 €
Each |
In Stock
Log in to see stock. |
||||||
Description
Choose the required Thermo Scientific™ Reverse Osmosis (RO) Membrane for your Thermo Scientific™ Barnstead™ point of use water purification system, as indicated.
Use Membranes with the Following Barnstead Systems:
09.2003:
- Reverse osmosis membrane with integrated pretreatment, 3L/hr for Smart2Pure™ systems
09.2006:
- Reverse osmosis membrane with integrated pretreatment, 6L/hr for Smart2Pure systems
22.0046:
- LabTower™ EDI 15 requires two of these RO membranes
- LabTower™ RO 20 requires two of these RO membranes
- LabTower RO 60 requires two of these RO membranes, along with two 22.0087 RO membranes
- LabTower™ TII 20 requires two of these RO membranes
- LabTower TII 60 requires two of these RO membranes, along with two 22.0087 RO membranes
- Pacific™ RO 3, 7, 12 require one of these RO membranes
- Pacific RO 20 requires two of these RO membranes
- Pacific™ TII 3, 7, 12 require one of these RO membranes
- Pacific TII 20 requires two of these RO membranes
22.0087:
- LabTower™ EDI 30 requires two of these RO membranes
- LabTower™ RO 40 requires two of these RO membranes
- LabTower RO 60 requires two of these RO membranes, along with two 22.0046 RO membranes
- LabTower TII 40 requires two of these RO membranes
- LabTower TII 60 requires two of these RO membranes, along with two 22.0046 RO membranes
- Pacific RO 40 requires two of these RO membranes
- Pacific TII 40 requires two of these RO membranes
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Reverse Osmosis Membranes