Learn More
RGX-104 hydrochloride, MedChemExpress
MedChemExpress RGX-104 hydrochloride is a small-molecule LXR agonist that modulates innate immunity via transcriptional activation of the ApoE gene.
Brand:  Medchem Express HY-111498
Encompass_Preferred
 View more versions of this product
Product Code. 16460787
  • 1242.00 € / 50mg
Explore more special offers
Description
Description
RGX-104 hydrochloride is a small-molecule LXR agonist that modulates innate immunity via transcriptional activation of the ApoE gene.
Specifications
Specifications
RGX-104 hydrochloride
White
C34H34Cl2F3NO3
DMSO : 150 mg/mL (237.14 mM; ultrasonic and warming and heat to 60°C)
632.54
99.14%
Solid
610318-03-1
Research
50 mg
O=C(O)CC1=CC=CC(OCC[C@H](N(CC2=CC=CC(C(F)(F)F)=C2Cl)CC(C3=CC=CC=C3)C4=CC=CC=C4)C)=C1.Cl
632.54
Research
COVID-19-immunoregulation