Learn More
RGX-104 hydrochloride, MedChemExpress
MedChemExpress RGX-104 hydrochloride is a small-molecule LXR agonist that modulates innate immunity via transcriptional activation of the ApoE gene.
160.65 € - 2118.15 €
Specifications
Chemical Name or Material | RGX-104 hydrochloride |
---|---|
CAS | 610318-03-1 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C34H34Cl2F3NO3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16485346
|
Medchem Express
HY-111498 |
100 mg |
2118.15 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16477197
|
Medchem Express
HY-111498 |
5 mg |
278.10 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16460787
|
Medchem Express
HY-111498 |
50 mg |
1242.00 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16450136
|
Medchem Express
HY-111498 |
2 mg |
160.65 €
2mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16434478
|
Medchem Express
HY-111498 |
10 mg |
423.90 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16433106
|
Medchem Express
HY-111498 |
25 mg |
847.80 €
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RGX-104 hydrochloride is a small-molecule LXR agonist that modulates innate immunity via transcriptional activation of the ApoE gene.Specifications
RGX-104 hydrochloride | |
White | |
C34H34Cl2F3NO3 | |
O=C(O)CC1=CC=CC(OCC[C@H](N(CC2=CC=CC(C(F)(F)F)=C2Cl)CC(C3=CC=CC=C3)C4=CC=CC=C4)C)=C1.Cl | |
632.54 | |
Research | |
COVID-19-immunoregulation |
610318-03-1 | |
Research | |
DMSO : 150 mg/mL (237.14 mM; ultrasonic and warming and heat to 60°C) | |
632.54 | |
99.14% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.