missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhenium(VI) oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Rhenium(VI) oxide, O3Re, CAS Number-1314-28-9, rhenium vi oxide trace metals basis 1g, rhenium vi oxide, rhenium trioxide, rhenia, rhenium oxide reo3, reo3, unii-fet0y2c413, 5g, 99.9% (metals basis), 400 deg.C (decomposition), 215-228-8, 236.2, YSZJKUDBYALHQE-UHFFFAOYSA-N, trioxorhenium
206.00 € - 1844.00 €
Chemical Identifiers
CAS | 1314-28-9 |
---|---|
Molecular Formula | O3Re |
Molecular Weight (g/mol) | 234.204 |
MDL Number | MFCD00016306 |
InChI Key | YSZJKUDBYALHQE-UHFFFAOYSA-N |
Synonym | rhenium trioxide, rhenium oxide reo3, rhenium vi oxide, unii-fet0y2c413, rhenia, reo3, rhenium vi oxide trace metals basis 1g |
PubChem CID | 102110 |
IUPAC Name | trioxorhenium |
SMILES | O=[Re](=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
11314779
|
Thermo Scientific Alfa Aesar
062109.03 |
1 g | |||||||
11304779
|
Thermo Scientific Alfa Aesar
062109.06 |
5 g |
913.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11346319
|
Thermo Scientific Alfa Aesar
062109.14 |
25 g |
1844.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
Rhenium(VI) oxide is involved as a catalyst for the hydrogenation of carboxylic acids and carboxamides. It is involved in the alkene epoxidation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificadores de productos químicos
1314-28-9 | |
234.204 | |
YSZJKUDBYALHQE-UHFFFAOYSA-N | |
102110 | |
O=[Re](=O)=O |
O3Re | |
MFCD00016306 | |
rhenium trioxide, rhenium oxide reo3, rhenium vi oxide, unii-fet0y2c413, rhenia, reo3, rhenium vi oxide trace metals basis 1g | |
trioxorhenium |
Especificaciones
400°C (decomposition) | |
Powder | |
O3Re | |
rhenium trioxide, rhenium oxide reo3, rhenium vi oxide, unii-fet0y2c413, rhenia, reo3, rhenium vi oxide trace metals basis 1g | |
YSZJKUDBYALHQE-UHFFFAOYSA-N | |
trioxorhenium | |
102110 | |
Moisture sensitive |
1314-28-9 | |
99.9% (metals basis) | |
MFCD00016306 | |
Insoluble in water. | |
O=[Re](=O)=O | |
234.204 | |
236.2 | |
Rhenium(VI) oxide |
Seguridad y manipulación
missing translation for 'einecsNumber' : 215-228-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only