missing translation for 'onlineSavingsMsg'
Learn More
Rhodanine, 98+%, Thermo Scientific Chemicals Product Code.: 11403847

Rhodanine, 98+%, Thermo Scientific Chemicals

Product Code. 11403847
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11403847

Brand: Thermo Scientific Alfa Aesar A18293.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rhodanine inhibits the multiplication of echovirus 12 and the development of virus-induced morphologic changes. It has been used in tannase assay in cultures of tannin degrading fungi. It possess anticonvulsant, antibacterial, antiviral and antidiabetic activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rhodanine inhibits the multiplication of echovirus 12 and the development of virus-induced morphologic changes. It has been used in tannase assay in cultures of tannin degrading fungi. It possess anticonvulsant, antibacterial, antiviral and antidiabetic activities.

Solubility
Soluble in water (2.25 g/L at 20°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-84-4
Molecular Formula C3H3NOS2
Molecular Weight (g/mol) 133.183
MDL Number MFCD00005488
InChI Key KIWUVOGUEXMXSV-UHFFFAOYSA-N
Synonym rhodanine, 2-thioxothiazolidin-4-one, 2-thioxo-4-thiazolidinone, rhodaninic acid, 4-thiazolidinone, 2-thioxo, 2-thioxo-1,3-thiazolidin-4-one, rhodanic acid, rhodanin, rodanin, 4-oxo-2-thioxothiazolineShow MoreShow Less
PubChem CID 1201546
ChEBI CHEBI:8830
IUPAC Name 2-sulfanylidene-1,3-thiazolidin-4-one
SMILES C1C(=O)NC(=S)S1

Specifications

Melting Point 166°C to 171°C
Density 0.868
Odor Odorless
Quantity 100 g
Beilstein 110701
Merck Index 14,8184
Solubility Information Soluble in water (2.25g/L at 20°C).
Formula Weight 133.19
Percent Purity ≥98%
Chemical Name or Material Rhodanine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberGBFA019684 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18293.14,A18293.36,A18293.22
Lot NumberJ08T046 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18293.14,A18293.36,A18293.22
Lot Number10141697 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18293.14,A18293.36,A18293.22
Lot Number10156461 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18293.14,A18293.36,A18293.22
Lot Number10177050 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18293.14,A18293.36,A18293.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rhodanine, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.