missing translation for 'onlineSavingsMsg'
Learn More

Rhodizonic acid disodium salt, 98%, Thermo Scientific Chemicals

41.20 € - 642.00 €

Chemical Identifiers

CAS 523-21-7
Molecular Formula C6Na2O6
Molecular Weight (g/mol) 214.04
MDL Number MFCD00001650
InChI Key SXWPCWBFJXDMAI-UHFFFAOYSA-L
Synonym sodium rhodizonate, sodium 3,4,5,6-tetraoxocyclohex-1-ene-1,2-bis olate, rhodizonic acid disodium salt, unii-9qf0329l3g, rhodizonic acid sodium salt, rhodizonic acid, disodium salt, disodium 3,4,5,6-tetraoxocyclohexene-1,2-diolate, 3,4,5,6-tetraoxocyclohexene-1,2-diol disodium salt, disodium tetraoxocyclohex-1-ene-1,2-bis olate, dipotassium tetraoxocyclohex-1-ene-1,2-bis olate
PubChem CID 68225
IUPAC Name disodium;3,4,5,6-tetraoxocyclohexene-1,2-diolate
SMILES [Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C(=O)C1=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11413167
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A16126.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11423167
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A16126.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11433167
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A16126.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Determining sulfate radical by barium chloride titration with rhodizonic acid disodium salt test paper as indicator has been established. Staining of thin sections for quick identification of quartz, potash feldspar and plagioclase is usually done with sodium hexanitrocobaltate (III) and rhodizonic acid disodium salt. N-allyl-N?-(sodium p-phenyl-sulfonate)thiourea is used for eliminating the interference of Ag +?Au 3+?Hg 2+in the identification of Sr 2+ using Rhodizonic acid disodium salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Determining sulfate radical by barium chloride titration with rhodizonic acid disodium salt test paper as indicator has been established. Staining of thin sections for quick identification of quartz, potash feldspar and plagioclase is usually done with sodium hexanitrocobaltate (III) and rhodizonic acid disodium salt. N-allyl-N′-(sodium p-phenyl-sulfonate)thiourea is used for eliminating the interference of Ag +?Au 3+?Hg 2+in the identification of Sr 2+ using Rhodizonic acid disodium salt.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

523-21-7
214.04
SXWPCWBFJXDMAI-UHFFFAOYSA-L
68225
[Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C(=O)C1=O
C6Na2O6
MFCD00001650
sodium rhodizonate, sodium 3,4,5,6-tetraoxocyclohex-1-ene-1,2-bis olate, rhodizonic acid disodium salt, unii-9qf0329l3g, rhodizonic acid sodium salt, rhodizonic acid, disodium salt, disodium 3,4,5,6-tetraoxocyclohexene-1,2-diolate, 3,4,5,6-tetraoxocyclohexene-1,2-diol disodium salt, disodium tetraoxocyclohex-1-ene-1,2-bis olate, dipotassium tetraoxocyclohex-1-ene-1,2-bis olate
disodium;3,4,5,6-tetraoxocyclohexene-1,2-diolate

Specifications

523-21-7
Odorless
MFCD00001650
14,8670
Soluble in water.
[Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C(=O)C1=O
214.04
214.04
Rhodizonic acid disodium salt
>300°C
C6Na2O6
5181379
sodium rhodizonate, sodium 3,4,5,6-tetraoxocyclohex-1-ene-1,2-bis olate, rhodizonic acid disodium salt, unii-9qf0329l3g, rhodizonic acid sodium salt, rhodizonic acid, disodium salt, disodium 3,4,5,6-tetraoxocyclohexene-1,2-diolate, 3,4,5,6-tetraoxocyclohexene-1,2-diol disodium salt, disodium tetraoxocyclohex-1-ene-1,2-bis olate, dipotassium tetraoxocyclohex-1-ene-1,2-bis olate
SXWPCWBFJXDMAI-UHFFFAOYSA-L
disodium;3,4,5,6-tetraoxocyclohexene-1,2-diolate
68225
98%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 208-340-3

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only